Product Details

SNP ID
rs200216272
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:112987054 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACTGGCAGGGTGGCTCCTATGGAC[A/G]GAAAACAGTCTTAAGAGGCAACTCC
Phenotype
MIM: 603350
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OAS2 PubMed Links

Gene Details

Gene
OAS2
Gene Name
2'-5'-oligoadenylate synthetase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001032731.1 334 Missense Mutation CAG,CGG Q65R NP_001027903.1
NM_002535.2 334 Missense Mutation CAG,CGG Q65R NP_002526.2
NM_016817.2 334 Missense Mutation CAG,CGG Q65R NP_058197.2

View Full Product Details