Product Details

SNP ID
rs199930976
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:56319341 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGATGACCCGGAAGTCCTTCTGCAG[A/G]CCATGACCCACAAACTTGACTCCAA
Phenotype
MIM: 605861
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CNPY2 PubMed Links

Gene Details

Gene
CNPY2
Gene Name
canopy FGF signaling regulator 2
There are no transcripts associated with this gene.

Gene
PAN2
Gene Name
PAN2 poly(A) specific ribonuclease subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001127460.2 3626 Silent Mutation GGC,GGT G1083G NP_001120932.1
NM_001166279.1 3626 Silent Mutation GGC,GGT G1082G NP_001159751.1
NM_014871.4 3626 Silent Mutation GGC,GGT G1079G NP_055686.3

View Full Product Details