Product Details
- SNP ID
-
rs199840248
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:52790268 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACTGCCTGAGCCGCCGCCCGCACTG[A/C]AGCCACCTCCTAAACCACCGCCCAT
- Phenotype
-
MIM: 148043
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
KRT3
PubMed Links
Gene Details
- Gene
- KRT3
- Gene Name
- keratin 3
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_057088.2 |
1727 |
Missense Mutation |
TGC,TTC |
C554F |
NP_476429.2 |
View Full Product Details