Product Details

SNP ID
rs202139017
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:57595890 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTGGCCTATTCTGTGTCCCCAGGT[A/G]TCCCTTACCCGAAACCCCCCCAGCG
Phenotype
MIM: 613142
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DTX3 PubMed Links

Gene Details

Gene
DTX3
Gene Name
deltex E3 ubiquitin ligase 3
There are no transcripts associated with this gene.

Gene
PIP4K2C
Gene Name
phosphatidylinositol-5-phosphate 4-kinase type 2 gamma
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001146258.1 496 Silent Mutation GTA,GTG V124V NP_001139730.1
NM_001146259.1 496 Silent Mutation GTA,GTG V106V NP_001139731.1
NM_001146260.1 496 Intron NP_001139732.1
NM_024779.4 496 Silent Mutation GTA,GTG V124V NP_079055.3
XM_005269152.2 496 Silent Mutation GTA,GTG V124V XP_005269209.1
XM_011538747.1 496 Silent Mutation GTA,GTG V124V XP_011537049.1
XM_017019973.1 496 UTR 5 XP_016875462.1

View Full Product Details