Product Details

SNP ID
rs200859761
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:52315161 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTGCAGCTGTACGCCACCCTGCTG[C/T]TGCTTCTCATTGTCTCGGTGCTCGC
Phenotype
MIM: 176804
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PTGER2 PubMed Links

Gene Details

Gene
PTGER2
Gene Name
prostaglandin E receptor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000956.3 864 Silent Mutation CTG,TTG L205L NP_000947.2

View Full Product Details