Product Details

SNP ID
rs201797824
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:64541015 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACCGAGCGCCTCATCGGCGACGCC[A/G]CCAAGAACCAGGTGGCCATGAACCC
Phenotype
MIM: 140560 MIM: 616578
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HSPA2 PubMed Links

Gene Details

Gene
HSPA2
Gene Name
heat shock protein family A (Hsp70) member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021979.3 548 Missense Mutation ACC,GCC T56A NP_068814.2
Gene
LOC102723809
Gene Name
uncharacterized LOC102723809
There are no transcripts associated with this gene.

Gene
PPP1R36
Gene Name
protein phosphatase 1 regulatory subunit 36
There are no transcripts associated with this gene.

Gene
ZBTB1
Gene Name
zinc finger and BTB domain containing 1
There are no transcripts associated with this gene.

View Full Product Details