Product Details
- SNP ID
-
rs201625686
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.14:88618273 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AATTCTGTCAATAGCGGCATGATCC[A/G]TAAGATGTTTTCCTGAAGGCACTTC
- Phenotype
-
MIM: 613279
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
EML5
PubMed Links
Gene Details
- Gene
- EML5
- Gene Name
- echinoderm microtubule associated protein like 5
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_183387.2 |
5532 |
Missense Mutation |
ACG,ATG |
T1866M |
NP_899243.1 |
XM_006720070.3 |
5532 |
Missense Mutation |
ACG,ATG |
T1858M |
XP_006720133.1 |
XM_011536528.2 |
5532 |
Missense Mutation |
ACG,ATG |
T1866M |
XP_011534830.1 |
XM_011536530.2 |
5532 |
Missense Mutation |
ACG,ATG |
T1863M |
XP_011534832.1 |
XM_011536531.2 |
5532 |
Missense Mutation |
ACG,ATG |
T1858M |
XP_011534833.1 |
XM_011536532.2 |
5532 |
Missense Mutation |
ACG,ATG |
T1858M |
XP_011534834.1 |
XM_011536533.2 |
5532 |
Missense Mutation |
ACG,ATG |
T1820M |
XP_011534835.1 |
XM_011536534.2 |
5532 |
Missense Mutation |
ACG,ATG |
T1807M |
XP_011534836.1 |
XM_011536536.2 |
5532 |
Intron |
|
|
XP_011534838.1 |
XM_011536538.2 |
5532 |
Intron |
|
|
XP_011534840.1 |
XM_017021062.1 |
5532 |
Missense Mutation |
ACG,ATG |
T1866M |
XP_016876551.1 |
XM_017021063.1 |
5532 |
Missense Mutation |
ACG,ATG |
T1859M |
XP_016876552.1 |
XM_017021064.1 |
5532 |
Missense Mutation |
ACG,ATG |
T1828M |
XP_016876553.1 |
XM_017021065.1 |
5532 |
Missense Mutation |
ACG,ATG |
T1851M |
XP_016876554.1 |
XM_017021066.1 |
5532 |
Missense Mutation |
ACG,ATG |
T1810M |
XP_016876555.1 |
XM_017021067.1 |
5532 |
Missense Mutation |
ACG,ATG |
T1809M |
XP_016876556.1 |
XM_017021068.1 |
5532 |
Missense Mutation |
ACG,ATG |
T1808M |
XP_016876557.1 |
XM_017021069.1 |
5532 |
Missense Mutation |
ACG,ATG |
T1807M |
XP_016876558.1 |
XM_017021070.1 |
5532 |
Intron |
|
|
XP_016876559.1 |
- Gene
- ZC3H14
- Gene Name
- zinc finger CCCH-type containing 14
There are no transcripts associated with this gene.
View Full Product Details