Product Details

SNP ID
rs200159674
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:52314581 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGCCTCCAATGACTCCCAGTCTGA[A/G]GACTGCGAGACGCGACAGTGGCTTC
Phenotype
MIM: 176804
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PTGER2 PubMed Links

Gene Details

Gene
PTGER2
Gene Name
prostaglandin E receptor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000956.3 284 Silent Mutation GAA,GAG E11E NP_000947.2

View Full Product Details