Product Details

SNP ID
rs202088808
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:105479183 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCGGCCCTGCCTGCGCTGCGAGCG[C/G]TGCGGGAAGACACTGACCCCCGGCG
Phenotype
MIM: 123875 MIM: 601183 MIM: 603526
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CRIP1 PubMed Links

Gene Details

Gene
CRIP1
Gene Name
cysteine rich protein 1
There are no transcripts associated with this gene.

Gene
CRIP2
Gene Name
cysteine rich protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001270837.1 997 Silent Mutation CGC,CGG R229R NP_001257766.1
NM_001270841.1 997 Silent Mutation CGC,CGG R34R NP_001257770.1
NM_001312.3 997 Silent Mutation CGC,CGG R155R NP_001303.1
Gene
MTA1
Gene Name
metastasis associated 1
There are no transcripts associated with this gene.

View Full Product Details