Product Details

SNP ID
rs199524651
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:43361239 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACGTAAGTCAGAACTCTTACTGAAA[C/T]TCTTTCCACAGTCAGGACACACATG
Phenotype
MIM: 609610
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ADAL PubMed Links

Gene Details

Gene
ADAL
Gene Name
adenosine deaminase like
There are no transcripts associated with this gene.

Gene
TUBGCP4
Gene Name
tubulin gamma complex associated protein 4
There are no transcripts associated with this gene.

Gene
ZSCAN29
Gene Name
zinc finger and SCAN domain containing 29
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152455.3 2980 Missense Mutation AAT,AGT N798S NP_689668.3
XM_006720401.3 2980 Missense Mutation AAT,AGT N798S XP_006720464.1
XM_011521266.2 2980 Missense Mutation AAT,AGT N798S XP_011519568.1
XM_017021949.1 2980 Intron XP_016877438.1

View Full Product Details