Product Details

SNP ID
rs201403716
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:67166346 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGAACGAGATCTTGTGGCGGGAGG[G/T]GGTGACACTTCGGCAGAGCCACGGT
Phenotype
MIM: 609077 MIM: 602438 MIM: 605235 MIM: 603500
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
FBXL8 PubMed Links

Gene Details

Gene
FBXL8
Gene Name
F-box and leucine rich repeat protein 8
There are no transcripts associated with this gene.

Gene
HSF4
Gene Name
heat shock transcription factor 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001040667.2 1477 Missense Mutation GGG,GTG G171V NP_001035757.1
NM_001538.3 1477 Missense Mutation GGG,GTG G171V NP_001529.2
Gene
KIAA0895L
Gene Name
KIAA0895 like
There are no transcripts associated with this gene.

Gene
NOL3
Gene Name
nucleolar protein 3
There are no transcripts associated with this gene.

Gene
TRADD
Gene Name
TNFRSF1A associated via death domain
There are no transcripts associated with this gene.

View Full Product Details