Product Details

SNP ID
rs200855023
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:67878263 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCTGGCAGATCTACATTGAGGGGC[A/G]AGATGAGCCAAGGTAAGGCAGGGCC
Phenotype
MIM: 606030 MIM: 605813
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
EDC4 PubMed Links

Gene Details

Gene
EDC4
Gene Name
enhancer of mRNA decapping 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014329.4 1231 Missense Mutation CAA,CGA Q331R NP_055144.3
Gene
NRN1L
Gene Name
neuritin 1 like
There are no transcripts associated with this gene.

Gene
NUTF2
Gene Name
nuclear transport factor 2
There are no transcripts associated with this gene.

View Full Product Details