Product Details

SNP ID
rs200001793
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:46884723 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTCTCCGCAAGCGCGCGATGCAGC[G/T]GGCGGCGGCGCTGGTCCGGCGGGGC
Phenotype
MIM: 138210
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
GPT2 PubMed Links

Gene Details

Gene
GPT2
Gene Name
glutamic--pyruvic transaminase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142466.2 118 Intron NP_001135938.1
NM_133443.3 118 Missense Mutation CGG,CTG R3L NP_597700.1
XM_017023790.1 118 Intron XP_016879279.1

View Full Product Details