Product Details

SNP ID
rs200926753
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:31459965 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTGTACGGAAGGGGCGTGCCGGAC[C/G]GAAGTGCGCAGACTCGGAGGCATAC
Phenotype
MIM: 615549
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ARMC5 PubMed Links

Gene Details

Gene
ARMC5
Gene Name
armadillo repeat containing 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001105247.1 1103 Silent Mutation ACC,ACG T147T NP_001098717.1
NM_001288767.1 1103 Silent Mutation ACC,ACG T242T NP_001275696.1
NM_001301820.1 1103 Silent Mutation ACC,ACG T179T NP_001288749.1
NM_024742.2 1103 Silent Mutation ACC,ACG T147T NP_079018.1
XM_006721091.2 1103 Silent Mutation ACC,ACG T179T XP_006721154.1

View Full Product Details