Product Details

SNP ID
rs201491122
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:1980032 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCGCAGCAGCACGTCCAGGCTCTCC[A/T]GGGCCTGCGCCTGAAAAGGCCTATC
Phenotype
MIM: 600924 MIM: 611256 MIM: 603927 MIM: 605915
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
GFER PubMed Links

Gene Details

Gene
GFER
Gene Name
growth factor, augmenter of liver regeneration
There are no transcripts associated with this gene.

Gene
NOXO1
Gene Name
NADPH oxidase organizer 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001267721.1 933 Missense Mutation CAG,CTG Q188L NP_001254650.1
NM_144603.3 933 Missense Mutation CAG,CTG Q183L NP_653204.1
NM_172167.2 933 Missense Mutation CAG,CTG Q184L NP_751907.1
NM_172168.2 933 Missense Mutation CAG,CTG Q189L NP_751908.1
XM_005255099.4 933 Missense Mutation CAG,CTG Q189L XP_005255156.1
XM_017022927.1 933 Missense Mutation CAG,CTG Q189L XP_016878416.1
XM_017022928.1 933 Missense Mutation CAG,CTG Q189L XP_016878417.1
Gene
SYNGR3
Gene Name
synaptogyrin 3
There are no transcripts associated with this gene.

Gene
TBL3
Gene Name
transducin beta like 3
There are no transcripts associated with this gene.

View Full Product Details