Product Details

SNP ID
rs201512959
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:46691011 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCAGGAAAGCAGAGGAGTACTTGC[A/G]CCTGTCCCGGGTGAAGTGTGTCGGC
Phenotype
MIM: 607213 MIM: 601501
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ORC6 PubMed Links

Gene Details

Gene
ORC6
Gene Name
origin recognition complex subunit 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014321.3 136 Missense Mutation CAC,CGC H29R NP_055136.1
XM_011522978.2 136 Missense Mutation CAC,CGC H29R XP_011521280.1
Gene
VPS35
Gene Name
VPS35 retromer complex component
There are no transcripts associated with this gene.

View Full Product Details