Product Details

SNP ID
rs202191694
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:30023775 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGTCAGCCCTGGGGACAGAGTCTC[C/T]GCGCTTCTTAGACAGACCTGACTTC
Phenotype
MIM: 604567 MIM: 615175
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C16orf92 PubMed Links

Gene Details

Gene
C16orf92
Gene Name
chromosome 16 open reading frame 92
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001109659.1 120 Missense Mutation CCG,CTG P38L NP_001103129.1
NM_001109660.1 120 Missense Mutation CCG,CTG P60L NP_001103130.1
Gene
DOC2A
Gene Name
double C2 domain alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282062.1 120 Intron NP_001268991.1
NM_001282063.1 120 Intron NP_001268992.1
NM_001282068.1 120 Intron NP_001268997.1
NM_003586.2 120 Intron NP_003577.2
XM_011545975.1 120 Intron XP_011544277.1
XM_017023776.1 120 Intron XP_016879265.1
XM_017023777.1 120 Intron XP_016879266.1
XM_017023778.1 120 Intron XP_016879267.1
Gene
FAM57B
Gene Name
family with sequence similarity 57 member B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318504.1 120 Intron NP_001305433.1
NM_031478.5 120 Intron NP_113666.2
XM_005255613.3 120 Intron XP_005255670.1
XM_005255614.3 120 Intron XP_005255671.1
XM_005255615.2 120 Intron XP_005255672.1
XM_017023749.1 120 Intron XP_016879238.1
XM_017023750.1 120 Intron XP_016879239.1
XM_017023751.1 120 Intron XP_016879240.1
XM_017023752.1 120 Intron XP_016879241.1
XM_017023753.1 120 Intron XP_016879242.1
XM_017023754.1 120 Intron XP_016879243.1

View Full Product Details