Product Details
- SNP ID
-
rs202104549
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:57154413 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CGCTGCTCCCAGAGTAGGCACCCAG[A/G]CCTGGGAGGATGCCGATACATACTG
- Phenotype
-
MIM: 604206
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CPNE2
PubMed Links
Gene Details
- Gene
- CPNE2
- Gene Name
- copine 2
There are no transcripts associated with this gene.
- Gene
- FAM192A
- Gene Name
- family with sequence similarity 192 member A
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_024946.2 |
901 |
Silent Mutation |
GGC,GGT |
G214G |
NP_079222.1 |
XM_005256156.4 |
901 |
Silent Mutation |
GGC,GGT |
G260G |
XP_005256213.1 |
XM_005256158.3 |
901 |
Silent Mutation |
GGC,GGT |
G220G |
XP_005256215.1 |
XM_005256160.2 |
901 |
Silent Mutation |
GGC,GGT |
G214G |
XP_005256217.1 |
XM_005256163.2 |
901 |
Silent Mutation |
GGC,GGT |
G214G |
XP_005256220.1 |
XM_005256164.3 |
901 |
Silent Mutation |
GGC,GGT |
G214G |
XP_005256221.1 |
XM_005256165.2 |
901 |
Silent Mutation |
GGC,GGT |
G214G |
XP_005256222.1 |
XM_006721275.3 |
901 |
Silent Mutation |
GGC,GGT |
G240G |
XP_006721338.2 |
XM_011523343.2 |
901 |
Silent Mutation |
GGC,GGT |
G214G |
XP_011521645.1 |
XM_017023681.1 |
901 |
Silent Mutation |
GGC,GGT |
G220G |
XP_016879170.1 |
XM_017023682.1 |
901 |
Silent Mutation |
GGC,GGT |
G214G |
XP_016879171.1 |
XM_017023683.1 |
901 |
Silent Mutation |
GGC,GGT |
G214G |
XP_016879172.1 |
XM_017023684.1 |
901 |
Silent Mutation |
GGC,GGT |
G214G |
XP_016879173.1 |
XM_017023685.1 |
901 |
Silent Mutation |
GGC,GGT |
G214G |
XP_016879174.1 |
XM_017023686.1 |
901 |
Silent Mutation |
GGC,GGT |
G214G |
XP_016879175.1 |
XM_017023687.1 |
901 |
Silent Mutation |
GGC,GGT |
G214G |
XP_016879176.1 |
XM_017023688.1 |
901 |
Silent Mutation |
GGC,GGT |
G214G |
XP_016879177.1 |
XM_017023689.1 |
901 |
Silent Mutation |
GGC,GGT |
G214G |
XP_016879178.1 |
XM_017023690.1 |
901 |
Silent Mutation |
GGC,GGT |
G214G |
XP_016879179.1 |
XM_017023691.1 |
901 |
Silent Mutation |
GGC,GGT |
G214G |
XP_016879180.1 |
XM_017023692.1 |
901 |
Silent Mutation |
GGC,GGT |
G214G |
XP_016879181.1 |
XM_017023693.1 |
901 |
Silent Mutation |
GGC,GGT |
G214G |
XP_016879182.1 |
XM_017023694.1 |
901 |
Silent Mutation |
GGC,GGT |
G214G |
XP_016879183.1 |
XM_017023695.1 |
901 |
Silent Mutation |
GGC,GGT |
G214G |
XP_016879184.1 |
XM_017023696.1 |
901 |
Silent Mutation |
GGC,GGT |
G214G |
XP_016879185.1 |
XM_017023697.1 |
901 |
Silent Mutation |
GGC,GGT |
G214G |
XP_016879186.1 |
XM_017023698.1 |
901 |
Silent Mutation |
GGC,GGT |
G214G |
XP_016879187.1 |
XM_017023699.1 |
901 |
Silent Mutation |
GGC,GGT |
G214G |
XP_016879188.1 |
XM_017023700.1 |
901 |
Silent Mutation |
GGC,GGT |
G214G |
XP_016879189.1 |
XM_017023701.1 |
901 |
Silent Mutation |
GGC,GGT |
G214G |
XP_016879190.1 |
XM_017023702.1 |
901 |
Silent Mutation |
GGC,GGT |
G214G |
XP_016879191.1 |
XM_017023703.1 |
901 |
Silent Mutation |
GGC,GGT |
G214G |
XP_016879192.1 |
XM_017023704.1 |
901 |
Silent Mutation |
GGC,GGT |
G214G |
XP_016879193.1 |
View Full Product Details