Product Details

SNP ID
rs200149460
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:41977301 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGCCAGGACCGCCAAAGAATGCCTT[A/G]AAGATATTGTTTGGATCAAAATCTG
Phenotype
MIM: 123830 MIM: 601964
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CNP PubMed Links

Gene Details

Gene
CNP
Gene Name
2',3'-cyclic nucleotide 3' phosphodiesterase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033133.4 1578 UTR 3 NP_149124.3
XM_006721701.3 1578 Intron XP_006721764.1
XM_011524340.2 1578 Intron XP_011522642.1
Gene
DNAJC7
Gene Name
DnaJ heat shock protein family (Hsp40) member C7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001144766.2 1578 Silent Mutation TTC,TTT F413F NP_001138238.1
NM_003315.3 1578 Silent Mutation TTC,TTT F469F NP_003306.3
XM_011525167.2 1578 Silent Mutation TTC,TTT F413F XP_011523469.1
XM_011525168.2 1578 Silent Mutation TTC,TTT F413F XP_011523470.1
XM_011525169.2 1578 Intron XP_011523471.1
XM_017024994.1 1578 Silent Mutation TTC,TTT F413F XP_016880483.1

View Full Product Details