Product Details

SNP ID
rs200286521
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:41842137 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGTTCATACTGCACAACTTTGAGG[A/C]GATGGTGAAAGTCTCGGCAGAATTT
Phenotype
MIM: 608778
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
KLHL10 PubMed Links

Gene Details

Gene
KLHL10
Gene Name
kelch like family member 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_011524706.2 784 Missense Mutation GAG,GCG E170A XP_011523008.1
Gene
KLHL11
Gene Name
kelch like family member 11
There are no transcripts associated with this gene.

Gene
NT5C3B
Gene Name
5'-nucleotidase, cytosolic IIIB
There are no transcripts associated with this gene.

View Full Product Details