Product Details

SNP ID
rs200478682
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:7042350 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCACGGCCACCACCAGCGCTGCTC[C/T]GTATCCCCCCAGGCCCCGGTCTAAA
Phenotype
MIM: 615765
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SLC16A11 PubMed Links

Gene Details

Gene
SLC16A11
Gene Name
solute carrier family 16 member 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153357.1 1403 Missense Mutation AGA,GGA R278G NP_699188.1
XM_005256488.3 1403 Missense Mutation AGA,GGA R373G XP_005256545.2
XM_017024281.1 1403 Missense Mutation AGA,GGA R373G XP_016879770.1
XM_017024282.1 1403 Intron XP_016879771.1
Gene
SLC16A13
Gene Name
solute carrier family 16 member 13
There are no transcripts associated with this gene.

View Full Product Details