Product Details

SNP ID
rs199648721
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:58169976 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCAGATCTTCTTCTTCCACTTTTT[A/G]GGAGGTGCCATGGTCTTCTTCCTCT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR4D2 PubMed Links

Gene Details

Gene
OR4D2
Gene Name
olfactory receptor family 4 subfamily D member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004707.3 321 Silent Mutation TTA,TTG L107L NP_001004707.1

View Full Product Details