Product Details

SNP ID
rs199823135
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:41989473 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGCCTGTACGAAAAACTGAACTGC[C/T]TTCTCAATACAATCTTCGTAATAAA
Phenotype
MIM: 601964
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DNAJC7 PubMed Links

Gene Details

Gene
DNAJC7
Gene Name
DnaJ heat shock protein family (Hsp40) member C7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001144766.2 855 Silent Mutation AAA,AAG K172K NP_001138238.1
NM_003315.3 855 Silent Mutation AAA,AAG K228K NP_003306.3
XM_011525167.2 855 Silent Mutation AAA,AAG K172K XP_011523469.1
XM_011525168.2 855 Silent Mutation AAA,AAG K172K XP_011523470.1
XM_011525169.2 855 Intron XP_011523471.1
XM_017024994.1 855 Silent Mutation AAA,AAG K172K XP_016880483.1

View Full Product Details