Product Details

SNP ID
rs200403711
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:38736150 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGTGTAGTATTCCTTCAGTGGCTC[C/G]TCCTCGTACAGAACCTCCACCTGGG
Phenotype
MIM: 611933 MIM: 600328 MIM: 600346
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CISD3 PubMed Links

Gene Details

Gene
CISD3
Gene Name
CDGSH iron sulfur domain 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001136498.1 1859 Intron NP_001129970.1
Gene
MLLT6
Gene Name
MLLT6, PHD finger domain containing
There are no transcripts associated with this gene.

Gene
PCGF2
Gene Name
polycomb group ring finger 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007144.2 1859 Silent Mutation GAC,GAG D199E NP_009075.1
XM_005257640.2 1859 Missense Mutation GAC,GAG D199E XP_005257697.1
XM_005257641.4 1859 Missense Mutation GAC,GAG D199E XP_005257698.1
XM_005257642.3 1859 Missense Mutation GAC,GAG D199E XP_005257699.1
XM_017025016.1 1859 Missense Mutation GAC,GAG D199E XP_016880505.1

View Full Product Details