Product Details

SNP ID
rs201917309
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:1716626 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGACACCGTCGTTCCCAGAACCCAG[C/T]GCGCTCTGTGAGTTGGCATTTTTAA
Phenotype
MIM: 612077 MIM: 614218
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MIR22 PubMed Links

Gene Details

Gene
MIR22
Gene Name
microRNA 22
There are no transcripts associated with this gene.

Gene
MIR22HG
Gene Name
MIR22 host gene
There are no transcripts associated with this gene.

Gene
TLCD2
Gene Name
TLC domain containing 2
There are no transcripts associated with this gene.

Gene
WDR81
Gene Name
WD repeat domain 81
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001163673.1 104 Silent Mutation AGC,AGT S17S NP_001157145.1
NM_001163809.1 104 Intron NP_001157281.1
NM_001163811.1 104 Intron NP_001157283.1
NM_152348.3 104 UTR 5 NP_689561.2
XM_011523651.2 104 Intron XP_011521953.1
XM_017024184.1 104 Intron XP_016879673.1

View Full Product Details