Product Details
- SNP ID
-
rs201138840
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:8476926 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCTCGCGGCTCAGGCCCTCGTTGGC[C/G]TCGGTGGCATCATCCAGTTCCCGCT
- Phenotype
-
MIM: 160776
MIM: 607538
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
MYH10
PubMed Links
Gene Details
- Gene
- MYH10
- Gene Name
- myosin, heavy chain 10, non-muscle
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001256012.1 |
5983 |
Missense Mutation |
GAC,GAG |
D1943E |
NP_001242941.1 |
NM_001256095.1 |
5983 |
Missense Mutation |
GAC,GAG |
D1921E |
NP_001243024.1 |
NM_005964.3 |
5983 |
Missense Mutation |
GAC,GAG |
D1912E |
NP_005955.3 |
XM_011523875.2 |
5983 |
Missense Mutation |
GAC,GAG |
D1973E |
XP_011522177.1 |
XM_011523878.2 |
5983 |
Missense Mutation |
GAC,GAG |
D1952E |
XP_011522180.1 |
XM_011523879.2 |
5983 |
Missense Mutation |
GAC,GAG |
D1951E |
XP_011522181.1 |
XM_011523880.2 |
5983 |
Missense Mutation |
GAC,GAG |
D1942E |
XP_011522182.1 |
XM_017024677.1 |
5983 |
Missense Mutation |
GAC,GAG |
D1922E |
XP_016880166.1 |
XM_017024678.1 |
5983 |
Missense Mutation |
GAC,GAG |
D1922E |
XP_016880167.1 |
XM_017024679.1 |
5983 |
Missense Mutation |
GAC,GAG |
D1922E |
XP_016880168.1 |
XM_017024680.1 |
5983 |
Missense Mutation |
GAC,GAG |
D1921E |
XP_016880169.1 |
XM_017024681.1 |
5983 |
Missense Mutation |
GAC,GAG |
D1912E |
XP_016880170.1 |
XM_017024682.1 |
5983 |
Missense Mutation |
GAC,GAG |
D1912E |
XP_016880171.1 |
- Gene
- NDEL1
- Gene Name
- nudE neurodevelopment protein 1 like 1
There are no transcripts associated with this gene.
View Full Product Details