Product Details

SNP ID
rs200982318
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:49702994 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGTCAGGGGACCAAAATACACAAC[C/T]GTCATAAAGATGAAGCTCTAGAGAA
Phenotype
MIM: 610790
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FAM117A PubMed Links

Gene Details

Gene
FAM117A
Gene Name
family with sequence similarity 117 member A
There are no transcripts associated with this gene.

Gene
SLC35B1
Gene Name
solute carrier family 35 member B1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278784.1 783 Silent Mutation ACA,ACG T193T NP_001265713.1
NM_005827.2 783 Silent Mutation ACA,ACG T297T NP_005818.2
XM_005256929.3 783 Silent Mutation ACA,ACG T193T XP_005256986.1
XM_006721632.3 783 Silent Mutation ACA,ACG T331T XP_006721695.1
XM_011524179.2 783 Silent Mutation ACA,ACG T254T XP_011522481.1
XM_011524180.2 783 Silent Mutation ACA,ACG T193T XP_011522482.1

View Full Product Details