Product Details

SNP ID
rs201733731
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:14301687 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTACTCGTGCGCCGGCTCCTGCGCC[G/T]CCGCGCCGGGGCTGCTGCTCCTGGG
Phenotype
MIM: 604058
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
HS3ST3B1 PubMed Links

Gene Details

Gene
HS3ST3B1
Gene Name
heparan sulfate-glucosamine 3-sulfotransferase 3B1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006041.2 638 Missense Mutation GCC,TCC A57S NP_006032.1
XM_017025479.1 638 Intron XP_016880968.1
Gene
MGC12916
Gene Name
uncharacterized protein MGC12916
There are no transcripts associated with this gene.

View Full Product Details