Product Details

SNP ID
rs200032614
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:39499342 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCTGGGCTCTCAGAGGAGGCCGCC[G/T]AGTCCCCGTGCGCCCTGGGCGCGGC
Phenotype
MIM: 602768 MIM: 607381
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
DLL3 PubMed Links

Gene Details

Gene
DLL3
Gene Name
delta like canonical Notch ligand 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016941.3 278 Nonsense Mutation GAG,TAG E74* NP_058637.1
NM_203486.2 278 Nonsense Mutation GAG,TAG E74* NP_982353.1
Gene
TIMM50
Gene Name
translocase of inner mitochondrial membrane 50
There are no transcripts associated with this gene.

View Full Product Details