Product Details

SNP ID
rs200086865
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:1112845 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTCGCCGCTCATGAAGCGCTGCTT[C/T]TCCCTGAGGTTCACGTGGTCGATGG
Phenotype
MIM: 138322 MIM: 615729
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GPX4 PubMed Links

Gene Details

Gene
GPX4
Gene Name
glutathione peroxidase 4
There are no transcripts associated with this gene.

Gene
SBNO2
Gene Name
strawberry notch homolog 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001100122.1 2590 Silent Mutation GAA,GAG E727E NP_001093592.1
NM_014963.2 2590 Silent Mutation GAA,GAG E784E NP_055778.2
XM_011527804.2 2590 Silent Mutation GAA,GAG E784E XP_011526106.1

View Full Product Details