Product Details

SNP ID
rs200154609
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:47260657 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCCCCAGGGGCCAGCAAGGGGGGC[C/T]GGACTCCTCCACAGCAGGGAGGCCG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CCDC9 PubMed Links

Gene Details

Gene
CCDC9
Gene Name
coiled-coil domain containing 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015603.2 487 Silent Mutation CGG,TGG R94W NP_056418.1
XM_017026578.1 487 Silent Mutation CGG,TGG R94W XP_016882067.1

View Full Product Details