Product Details

SNP ID
rs201263017
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:5587254 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAGCGAGTGGGACTTAGTAGCGGC[G/T]GGTGAAGTGGGGGTACGGGGGGGGA
Phenotype
MIM: 608066
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
SAFB2 PubMed Links

Gene Details

Gene
SAFB2
Gene Name
scaffold attachment factor B2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014649.2 2413 Missense Mutation GGC,TGC G951C NP_055464.1
XM_011528449.2 2413 UTR 3 XP_011526751.1
XM_011528450.2 2413 Missense Mutation GGC,TGC G497C XP_011526752.1
XM_017027520.1 2413 Missense Mutation GGC,TGC G497C XP_016883009.1
XM_017027521.1 2413 Missense Mutation GGC,TGC G497C XP_016883010.1
XM_017027522.1 2413 Missense Mutation GGC,TGC G497C XP_016883011.1

View Full Product Details