Product Details

SNP ID
rs200713204
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:47260651 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCTCGGCCCCCAGGGGCCAGCAAG[C/G]GGGGCCGGACTCCTCCACAGCAGGG
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CCDC9 PubMed Links

Gene Details

Gene
CCDC9
Gene Name
coiled-coil domain containing 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015603.2 481 Missense Mutation CGG,GGG R92G NP_056418.1
XM_017026578.1 481 Missense Mutation CGG,GGG R92G XP_016882067.1

View Full Product Details