Product Details

SNP ID
rs201922550
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:32598331 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTTCTCAGCAGCATGAGATCCACT[C/G]TGAGCTGGCAGGTTATTCTGTCTCA
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ANKRD27 PubMed Links

Gene Details

Gene
ANKRD27
Gene Name
ankyrin repeat domain 27
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032139.2 3123 Missense Mutation CAC,CAG H989Q NP_115515.2
XM_006723412.2 3123 Missense Mutation CAC,CAG H989Q XP_006723475.1
XM_017027349.1 3123 Missense Mutation CAC,CAG H793Q XP_016882838.1

View Full Product Details