Product Details

SNP ID
rs202205669
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:17807829 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGGATGAAGTATCTCCGGCACCGG[C/T]GGCCCAATGCCACCCTCATTCTGGC
Phenotype
MIM: 605863 MIM: 146738
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
B3GNT3 PubMed Links

Gene Details

Gene
B3GNT3
Gene Name
UDP-GlcNAc:betaGal beta-1,3-N-acetylglucosaminyltransferase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014256.3 169 Missense Mutation CGG,TGG R8W NP_055071.2
XM_011527626.2 169 Missense Mutation CGG,TGG R8W XP_011525928.1
Gene
INSL3
Gene Name
insulin like 3
There are no transcripts associated with this gene.

View Full Product Details