Product Details

SNP ID
rs201822855
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:4504876 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAGACCCACCTCCTGGACAGCAGC[A/G]TCCTCCGCACTGGCACCTGAGCCCT
Phenotype
MIM: 613247
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HDGFRP2 PubMed Links

Gene Details

Gene
HDGFRP2
Gene Name
hepatoma-derived growth factor-related protein 2
There are no transcripts associated with this gene.

Gene
PLIN4
Gene Name
perilipin 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080400.1 5693 Silent Mutation GAC,GAT D1244D NP_001073869.1
XM_006722866.2 5693 Silent Mutation GAC,GAT D1259D XP_006722929.1
XM_006722868.3 5693 Silent Mutation GAC,GAT D1258D XP_006722931.1
XM_011528233.2 5693 Silent Mutation GAC,GAT D1319D XP_011526535.1
XM_017027192.1 5693 Silent Mutation GAC,GAT D1320D XP_016882681.1
XM_017027193.1 5693 Silent Mutation GAC,GAT D1320D XP_016882682.1
XM_017027194.1 5693 Silent Mutation GAC,GAT D1320D XP_016882683.1

View Full Product Details