Product Details
- SNP ID
-
rs201142610
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:45614619 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCAGAACTCACCAAACACCCCAAAC[C/T]CTAGGACACAAGTAGCTGTGGCCCA
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
EML2
PubMed Links
Gene Details
- Gene
- EML2
- Gene Name
- echinoderm microtubule associated protein like 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001193268.1 |
2436 |
Missense Mutation |
GAG,GGG |
E761G |
NP_001180197.1 |
NM_001193269.1 |
2436 |
Missense Mutation |
GAG,GGG |
E707G |
NP_001180198.1 |
NM_012155.2 |
2436 |
Missense Mutation |
GAG,GGG |
E560G |
NP_036287.1 |
XM_006723125.2 |
2436 |
Missense Mutation |
GAG,GGG |
E760G |
XP_006723188.1 |
XM_006723126.2 |
2436 |
Missense Mutation |
GAG,GGG |
E706G |
XP_006723189.1 |
XM_011526694.2 |
2436 |
Missense Mutation |
GAG,GGG |
E729G |
XP_011524996.1 |
XM_011526695.2 |
2436 |
Missense Mutation |
GAG,GGG |
E716G |
XP_011524997.1 |
XM_011526698.2 |
2436 |
Missense Mutation |
GAG,GGG |
E538G |
XP_011525000.1 |
XM_011526700.2 |
2436 |
Missense Mutation |
GAG,GGG |
E444G |
XP_011525002.1 |
XM_011526701.2 |
2436 |
Missense Mutation |
GAG,GGG |
E375G |
XP_011525003.1 |
XM_017026550.1 |
2436 |
Missense Mutation |
GAG,GGG |
E528G |
XP_016882039.1 |
XM_017026551.1 |
2436 |
Intron |
|
|
XP_016882040.1 |
View Full Product Details