Product Details

SNP ID
rs201177605
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:1108855 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCTGCTGCTGACGTCGGCCATGAC[A/G]GCGGCGATGCGGCCCCACACGCGCA
Phenotype
MIM: 138322 MIM: 615729
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GPX4 PubMed Links

Gene Details

Gene
GPX4
Gene Name
glutathione peroxidase 4
There are no transcripts associated with this gene.

Gene
SBNO2
Gene Name
strawberry notch homolog 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001100122.1 3778 Silent Mutation GCC,GCT A1123A NP_001093592.1
NM_014963.2 3778 Silent Mutation GCC,GCT A1180A NP_055778.2
XM_011527804.2 3778 Silent Mutation GCC,GCT A1180A XP_011526106.1

View Full Product Details