Product Details

SNP ID
rs201010975
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:48703047 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCCACTATATTTCACGTTCAGCAG[C/T]GGCTAGCGAAGATTCAAGCCATGTG
Phenotype
MIM: 182100 MIM: 610349
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FUT2 PubMed Links

Gene Details

Gene
FUT2
Gene Name
fucosyltransferase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000511.5 Intron NP_000502.4
NM_001097638.2 Intron NP_001091107.1
Gene
LOC105447645
Gene Name
uncharacterized LOC105447645
There are no transcripts associated with this gene.

Gene
MAMSTR
Gene Name
MEF2 activating motif and SAP domain containing transcriptional regulator
There are no transcripts associated with this gene.

View Full Product Details