Product Details

SNP ID
rs61752096
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:2406515 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTGCACCACGCGGTGATGCACTCC[T/C]AGCAGAACAGGTGGCCGCAGGGCGT
Phenotype
MIM: 602859 MIM: 612836
Polymorphism
T/C, Transition substitution
Allele Nomenclature
Literature Links
PEX10 PubMed Links

Gene Details

Gene
PEX10
Gene Name
peroxisomal biogenesis factor 10
There are no transcripts associated with this gene.

Gene
PLCH2
Gene Name
phospholipase C eta 2
There are no transcripts associated with this gene.

Gene
RER1
Gene Name
retention in endoplasmic reticulum sorting receptor 1
There are no transcripts associated with this gene.

View Full Product Details