Product Details

SNP ID
rs201709727
Assay Type
Functionally tested
NCBI dbSNP Submissions
6
Location
Chr.1:159535487 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGGTAAATGTCACAAAAGAAGTGG[G/T]CTACCACTGTGCCACAGAACGGCAA
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
OR10J5 PubMed Links

Gene Details

Gene
OR10J5
Gene Name
olfactory receptor family 10 subfamily J member 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004469.1 521 Missense Mutation GAC,GCC D174A NP_001004469.1

View Full Product Details