Product Details

SNP ID
rs201088681
Assay Type
Functionally tested
NCBI dbSNP Submissions
6
Location
Chr.1:26280871 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACGCCCTGAGGGATGAGATGCGAG[C/T]CTTGGCAGGCAACCCCAAGGCCACC
Phenotype
MIM: 615679 MIM: 609151
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CEP85 PubMed Links

Gene Details

Gene
CEP85
Gene Name
centrosomal protein 85
There are no transcripts associated with this gene.

Gene
SH3BGRL3
Gene Name
SH3 domain binding glutamate rich protein like 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_031286.3 589 Missense Mutation GCC,GTC A52V NP_112576.1
Gene
UBXN11
Gene Name
UBX domain protein 11
There are no transcripts associated with this gene.

View Full Product Details