Product Details

SNP ID
rs201490344
Assay Type
Functionally tested
NCBI dbSNP Submissions
9
Location
Chr.1:206866568 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAAAAACTACCAGACCCCTGACCAT[C/T]ATACTCTCCGGAAGATCAGCAGCCT
Phenotype
MIM: 605619
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IL20 PubMed Links

Gene Details

Gene
IL20
Gene Name
interleukin 20
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018724.3 780 Missense Mutation CAT,TAT H104Y NP_061194.2
XM_005273152.3 780 Missense Mutation CAT,TAT H189Y XP_005273209.1

View Full Product Details