Product Details

SNP ID
rs200764024
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:25245208 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCACGGCTTGTTTCTTTGGCTGAA[G/T]GAACAGTTCTGAGACTAGCTGGCAA
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
RSRP1 PubMed Links

Gene Details

Gene
RSRP1
Gene Name
arginine and serine rich protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321772.1 856 Missense Mutation CAT,CCT H205P NP_001308701.1
NM_020317.4 856 Missense Mutation CAT,CCT H205P NP_064713.3
XM_011541797.1 856 Missense Mutation CAT,CCT H205P XP_011540099.1

View Full Product Details