Product Details

SNP ID
rs200221315
Assay Type
Functionally tested
NCBI dbSNP Submissions
5
Location
Chr.1:54200388 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCGGAGCCTCCTCCCCTGGATAGG[G/T]TGTACGAGATCCCTGGACTGGAGCC
Phenotype
MIM: 611843
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CYB5RL PubMed Links

Gene Details

Gene
CYB5RL
Gene Name
cytochrome b5 reductase like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001031672.2 222 Intron NP_001026842.2
Gene
MRPL37
Gene Name
mitochondrial ribosomal protein L37
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016491.3 222 Missense Mutation GTG,TTG V49L NP_057575.2
XM_005270920.3 222 Missense Mutation GTG,TTG V49L XP_005270977.1

View Full Product Details