Product Details

SNP ID
rs200729328
Assay Type
Functionally tested
NCBI dbSNP Submissions
11
Location
Chr.1:154569495 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTACGGATACAGAGGAGCGGCTGG[G/T]GGAGCATCTCCTGGATCCTTCCCGC
Phenotype
MIM: 118507
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CHRNB2 PubMed Links

Gene Details

Gene
CHRNB2
Gene Name
cholinergic receptor nicotinic beta 2 subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000748.2 344 Missense Mutation GGG,GTG G33V NP_000739.1
XM_017000180.1 344 UTR 5 XP_016855669.1

View Full Product Details