Product Details

SNP ID
rs200919340
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:145912174 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTACCGGAAGGTCAGGGCTTGAGT[T/C]GTAGCCAGGGATAGATTAGGGTGAG
Phenotype
MIM: 612875 MIM: 604042 MIM: 603867 MIM: 605313
Polymorphism
T/C, Transition substitution
Allele Nomenclature
Literature Links
GNRHR2 PubMed Links

Gene Details

Gene
GNRHR2
Gene Name
gonadotropin releasing hormone receptor 2 (pseudogene)
There are no transcripts associated with this gene.

Gene
ITGA10
Gene Name
integrin subunit alpha 10
There are no transcripts associated with this gene.

Gene
PEX11B
Gene Name
peroxisomal biogenesis factor 11 beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001184795.1 809 Missense Mutation NP_001171724.1
NM_003846.2 809 Missense Mutation NP_003837.1
Gene
RBM8A
Gene Name
RNA binding motif protein 8A
There are no transcripts associated with this gene.

View Full Product Details