Product Details
- SNP ID
-
rs200212806
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
3
- Location
-
Chr.1:119422221 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACATTCTGGCCTTGAGGGCTCTGCG[A/G]GACCCCAAGAAGGCCCCAAGTGTCC
- Phenotype
-
MIM: 613890
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
HSD3B2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs33926546] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- HSD3B2
- Gene Name
- hydroxy-delta-5-steroid dehydrogenase, 3 beta- and steroid delta-isomerase 2
View Full Product Details