Product Details
- SNP ID
-
rs200227851
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
3
- Location
-
Chr.1:149943890 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGTGCACCAGGAGCTCCCCATCCGG[G/T]TCCCGCTCCCTCCTCCTCAGTTCTT
- Phenotype
-
MIM: 611748
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
MTMR11
PubMed Links
Gene Details
- Gene
- MTMR11
- Gene Name
- myotubularin related protein 11
There are no transcripts associated with this gene.
- Gene
- OTUD7B
- Gene Name
- OTU deubiquitinase 7B
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_020205.3 |
2597 |
Missense Mutation |
GAA,GAC |
E833D |
NP_064590.2 |
XM_011509782.1 |
2597 |
Missense Mutation |
GAA,GAC |
E842D |
XP_011508084.1 |
XM_011509783.2 |
2597 |
Missense Mutation |
GAA,GAC |
E842D |
XP_011508085.1 |
XM_011509784.2 |
2597 |
Missense Mutation |
GAA,GAC |
E842D |
XP_011508086.1 |
XM_011509785.2 |
2597 |
Missense Mutation |
GAA,GAC |
E842D |
XP_011508087.1 |
XM_011509786.1 |
2597 |
Missense Mutation |
GAA,GAC |
E763D |
XP_011508088.1 |
XM_011509787.1 |
2597 |
Missense Mutation |
GAA,GAC |
E763D |
XP_011508089.1 |
XM_011509788.1 |
2597 |
Missense Mutation |
GAA,GAC |
E763D |
XP_011508090.1 |
XM_017001850.1 |
2597 |
Missense Mutation |
GAA,GAC |
E754D |
XP_016857339.1 |
XM_017001851.1 |
2597 |
Missense Mutation |
GAA,GAC |
E704D |
XP_016857340.1 |
View Full Product Details