Product Details

SNP ID
rs200190364
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:183252719 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTGGGACAGGTAATCCACAACATG[A/G]CCGTCCCCATGCTGCAGGGCCAGCC
Phenotype
MIM: 150292 MIM: 608701
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LAMC2 PubMed Links

Gene Details

Gene
LAMC2
Gene Name
laminin subunit gamma 2
There are no transcripts associated with this gene.

Gene
NMNAT2
Gene Name
nicotinamide nucleotide adenylyltransferase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015039.3 1078 Silent Mutation GGC,GGT G282G NP_055854.1
NM_170706.3 1078 Silent Mutation GGC,GGT G277G NP_733820.1

View Full Product Details